Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol red | 143-74-8 | 98.0% | 25 G
SDP

Catalog No. 5000414203
Encompass_Preferred

Phenol red (Phenolsulfonephthalein) is a pH indicator dye that displays a distinct color change from pink to yellow in the event of a positive reaction. It is commonly used as a standard pH indicator in various cell and tissue culture media to quickly check the health of the culture. Additionally, it has been utilized in protocols for detecting cellular hydrogen peroxide and peroxidase activity from human peroxidase enzymes.

  • pH indicator dye
  • Color change from pink to yellow
  • Used in cell and tissue culture media
  • Can detect cellular hydrogen peroxide and peroxidase activity
  • Solid appearance, red to reddish brown color
  • For research use only

Catalog No. 50-004-14203 Supplier Medchemexpress LLC Supplier No. 25GHYD0169
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenol red (Phenolsulfonephthalein) is a pH indicator dye that displays a distinct color change from pink to yellow in the event of a positive reaction. It is commonly used as a standard pH indicator in various cell and tissue culture media to quickly check the health of the culture. Additionally, it has been utilized in protocols for detecting cellular hydrogen peroxide and peroxidase activity from human peroxidase enzymes.

  • pH indicator dye
  • Color change from pink to yellow
  • Used in cell and tissue culture media
  • Can detect cellular hydrogen peroxide and peroxidase activity
  • Solid appearance, red to reddish brown color
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.