Learn More
ATCC Phenol Red
Catalog No. 502384156
Encompass
Phenol red can be used in cell culture media as a pH indicator. It is provided as an optional media component when preparing complete media as a part of the complete ATCC Primary Cell Solutions system.
Catalog No. 50-238-4156 Supplier ATCC Supplier No. PCS999001
Phenol red can be used in cell culture media as a pH indicator. It is provided as an optional media component when preparing complete media as a part of the complete ATCC Primary Cell Solutions system.