Learn More
Description
Phenol Red is a phenolsulfonphthalein compound. It is present as a sulfonated hydroxyquinone in weakly acidic aqueous solutions. In alkaline solutions, the dye forms a hydrophilic dianion (a small, red anion and has a larger conjugated system than the yellow hydroxyquinone).
Specifications
Specifications
| CAS | 143-74-8 |
| Molecular Weight (g/mol) | 354.38 g/mol |
| Color | Red to Rust |
| Quantity | 25 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.