Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phenol Red Broth w/1% Lactose

Catalog No. R062326 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 5 mL
20 x 7 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R062326 20 x 7 mL
R062322 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R062326 Supplier Thermo Scientific™ Supplier No. R062326
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Determine fermentation reactions of bacteria using Phenol Red Broth with 1% Lactose.

Determine fermentation reactions of bacteria with Thermo Scientific™ Remel™ Phenol Red Broth with 1% Lactose.

Vera recommended using casein peptone in fermentation test media because of its high growth-promoting qualities1. It was reported that peptone could be used with phenol red as the pH indicator in fermentation tests.

Phenol Red Broth is a complete basal medium prepared with phenol red as an indicator, which changes the medium to yellow when acid is produced. Carbohydrates may be added to the broth base in a final concentration of 1%. A Phenol Red Broth Base Control

TRUSTED_SUSTAINABILITY

Specifications

Description Phenol Red Broth w/1% Lactose
Format 15mm x 103mm Durham Tube
Product Type Microbiology Media
Quantity 20 x 7 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.