Learn More
Bioworld Phenol Red Broth Base, 500 g
Catalog No. 50254354
Encompass_Preferred
Phenol Red Broth BaseUsed with the addition of carbohydrates for the differentiation of microorganisms by their fermentation reactionsCan be supplemented with carbohydratespH: 7.4Working Concentration: 15 g/LDehydrated Appearance: light reddish to white, homogeneous powderPrepared Appearance: bright red to red-orange broth (appearance can change with addition of carbohydrates)Soluble in waterComposition (g/L):Pancreatic Digest of Casein: 10gSodium Chlorid: 5g Phenol Red: 0.018g
Catalog No. 50-254-354 Supplier Bioworld Supplier No. 306200651
Phenol Red Broth BaseUsed with the addition of carbohydrates for the differentiation of microorganisms by their fermentation reactionsCan be supplemented with carbohydratespH: 7.4Working Concentration: 15 g/LDehydrated Appearance: light reddish to white, homogeneous powderPrepared Appearance: bright red to red-orange broth (appearance can change with addition of carbohydrates)Soluble in waterComposition (g/L):Pancreatic Digest of Casein: 10gSodium Chlorid: 5g Phenol Red: 0.018g