Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phenol Red Broth with 1% Dextrose and Durham Tube

Catalog No. R062242 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R062242 20 x 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R062242 Supplier Thermo Scientific™ Supplier No. R062242
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Determine fermentation reactions of bacteria using Phenol Red Broth w/1% Dextrose.

Determine fermentation reactions of bacteria with Thermo Scientific™ Remel™ Phenol Red Broth w/1% Dextrose.

Vera recommended using casein peptone in fermentation test media because of its high growth-promoting qualities1. It was reported that peptone could be used with phenol red as the pH indicator in fermentation tests.

Phenol Red Broth is a complete basal medium prepared with phenol red as an indicator, which changes the medium to yellow when acid is produced. Carbohydrates may be added to the broth base in a final concentration of 1%. A Phenol Red Broth Base Control tube is used as a negative control for fermentation studies. A Durham tube may be inserted in a tube of Phenol Red Broth w/1% Dextrose to allow detection of gas production.

  • Can be used in fermentation studies of pure culture of many species of bacteria due to high growth promoting qualities of casein peptone.
  • Recommended by Association of Official Analytical Collaboration (AOAC) International and the American Public Health Association (APHA)2-5.
  • Easy to interpret because of fermentation reaction and gas production. Yellow color is observed when there is acid production and red-orange color is observed when there is no fermentation. Bubbles are produced in the Durham tube when gas is produced and no bubbles is produced in the Durham tube when there is no gas production.

Phenol Red Broth w/1% Dextrose contains casein peptone that has high growth promoting qualities. Phenol red is the indicator which helps to determine the reaction by color change.

This medium also contains Sodium Chloride, Phenol Red, Demineralized Water and Dextrose. Sodium Chloride supplies essential electrolytes and maintains osmotic equilibrium of the medium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Phenol Red Broth w/1% Dextrose
Format 15mm x 103mm Durham Tube
Product Type Microbiology Media
Quantity 20 x 7 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.