Learn More
Hach Company Phenol Red Indicator Solution, 100 mL MDB
Supplier:  Hach Company 21132
Encompass
pH Indicator Solution. Color range: pH 6.5-8.5. 100 mL Marked Dropping Bottle
Catalog No. 50-199-4210