Learn More
Medchemexpress LLC HY-D0169A 5g Medchemexpress, Phenol Red (sodium salt) CAS:34487-61-1 Purity:>98%
SDP
Catalog No. 501933152
Encompass_Preferred
Medchemexpress, HY-D0169A 5g Phenol Red (sodium salt) CAS:34487-61-1 Phenol Red sodium salt is a water soluble pH indicator used in the 6.8 (yellow) to 8.2 (red) range. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3152 Supplier Medchemexpress LLC Supplier No. HYD0169A5G
Medchemexpress, HY-D0169A 5g Phenol Red (sodium salt) CAS:34487-61-1 Phenol Red sodium salt is a water soluble pH indicator used in the 6.8 (yellow) to 8.2 (red) range. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.