Learn More
Abcam Phenol red, pH indicator, 25G
Catalog No. 502704707
Encompass_Preferred
MW 354.37 g/mol. pH indicator widely used in culture media to identify pH changes from neutral (red) to acidic (yellow) values. Typically a change in color from red to yellow indicates high rates of cellular death when used in tissue cultures. Observed to act as a weak estrogen in the culture of breast cancer cells. Induced oogenesis from cells scraped from the surface of adult ovaries.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4707 Supplier Abcam Supplier No. AB14633625G
MW 354.37 g/mol. pH indicator widely used in culture media to identify pH changes from neutral (red) to acidic (yellow) values. Typically a change in color from red to yellow indicates high rates of cellular death when used in tissue cultures. Observed to act as a weak estrogen in the culture of breast cancer cells. Induced oogenesis from cells scraped from the surface of adult ovaries.
The product is subject to the following: Abcam Restricted Use Statement