Learn More
Crescent Chemical Co Inc PHENOL RED RESEARCH GRADE
SDP
Catalog No. 50248003
Encompass_Preferred
Phenol Red research grade Indicator pH 6.5 - 8.0. Tested for use in tissue culture. Size - 5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 32095.01 CAS 143-74-8
Catalog No. 50-248-003 Supplier Crescent Chemical Co Inc Supplier No. SE32095.01
Phenol Red research grade Indicator pH 6.5 - 8.0. Tested for use in tissue culture. Size - 5G Storage Conditions - +15 °C TO +30 °C Catalog Number - 32095.01 CAS 143-74-8