Learn More
Apexbio Technology LLC Phenol Red sodium salt 34487-61-1 5g
SDP
Catalog No. 5000093467
Encompass_Preferred
Phenol Red sodium salt (34487-61-1) is a small molecule compound binding to estrogen receptors It can interact with estrogen receptors thereby modulating downstream signaling pathways and associated cellular processes Phenol Red sodium salt functions primarily via estrogen receptor affinity In cell-based studies it binds to estrogen receptors in human breast cancer cell lines such as MCF-7 cells Based on these properties Phenol Red sodium salt holds research potential in oncology studies particularly those investigating hormone-mediated signaling pathways and receptor-mediated cellular responses
Catalog No. 50-000-93467 Supplier Apexbio Technology LLC Supplier No. A47165000
Phenol Red sodium salt (34487-61-1) is a small molecule compound binding to estrogen receptors It can interact with estrogen receptors thereby modulating downstream signaling pathways and associated cellular processes Phenol Red sodium salt functions primarily via estrogen receptor affinity In cell-based studies it binds to estrogen receptors in human breast cancer cell lines such as MCF-7 cells Based on these properties Phenol Red sodium salt holds research potential in oncology studies particularly those investigating hormone-mediated signaling pathways and receptor-mediated cellular responses