Learn More
Medchemexpress LLC Phenol red (sodium salt) | 34487-61-1 | 97.0% | 25 G
SDP
Catalog No. 5000413933
Encompass_Preferred
Phenol Red (Phenolsulfonephthalein) sodium salt is a pH indicator dye. It also acts as an antagonist of rat P2X1R and enhances the halogenating activity of myeloperoxidase. This compound exhibits a distinct color change from pink to yellow in a positive pH reaction.
  • Ph indicator dye
  • Antagonist of rat P2X1R
  • Enhances the halogenating activity of myeloperoxidase
  • Shows a distinct color change from pink to yellow in a positive pH reaction
Catalog No. 50-004-13933 Supplier Medchemexpress LLC Supplier No. 25GHYD0169A
Phenol Red (Phenolsulfonephthalein) sodium salt is a pH indicator dye. It also acts as an antagonist of rat P2X1R and enhances the halogenating activity of myeloperoxidase. This compound exhibits a distinct color change from pink to yellow in a positive pH reaction.
  • Ph indicator dye
  • Antagonist of rat P2X1R
  • Enhances the halogenating activity of myeloperoxidase
  • Shows a distinct color change from pink to yellow in a positive pH reaction