Learn More
Selleck Chemical LLC Phenol Red sodium salt S4418-100mg
SDP
Catalog No. 503110245
Encompass_Preferred
Phenol Red sodium salt (Phenolsulfonephthalein sodium salt) is used ubiquitously as a pH indicator in tissue culture media ranging from 6 8 (yellow) to 8 2 (red) Phenol red binds to the estrogen receptor of MCF-7 human breast cancer cells
Catalog No. 50-311-0245 Supplier Selleck Chemical LLC Supplier No. S4418100MG
Phenol Red sodium salt (Phenolsulfonephthalein sodium salt) is used ubiquitously as a pH indicator in tissue culture media ranging from 6 8 (yellow) to 8 2 (red) Phenol red binds to the estrogen receptor of MCF-7 human breast cancer cells