Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Phenol Red, Sodium Salt, 6.8-8.2 pH 5 GR
SDP

Catalog No. 5027839
Encompass_Preferred

(Phenolsulfonphthalein Sodium Salt)
CAS #34487-61-1 C₁₉H₁₃NaO₅S F.W. 376.37
Assay: >99.0%

Also known as Phenolsulfonphthalein sodium salt. A standard indicator for preparing buffers, pH range 6.8-8.2.
pH: 6.8-8.2

Catalog No. 50-278-39 Supplier Electron Microscopy Sciences Supplier No. 19328
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Phenolsulfonphthalein Sodium Salt)
CAS #34487-61-1 C₁₉H₁₃NaO₅S F.W. 376.37
Assay: >99.0%

Also known as Phenolsulfonphthalein sodium salt. A standard indicator for preparing buffers, pH range 6.8-8.2.
pH: 6.8-8.2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.