Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol Red Sodium Salt, ACS Reagent (Phenolsulfonphthalein Sodium Salt), Reagents

Catalog No. 01336730
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-730 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-730 Supplier Reagents Holdings Llc Supplier No. C61226005C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenol Red Sodium Salt, ACS Reagent (Phenolsulfonphthalein Sodium Salt), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Phenol Red Sodium Salt, ACS Reagent (Phenolsulfonphthalein Sodium Salt)
Boiling Point 285°C
CAS 34487-61-1
Packaging Glass Bottle
Specific Gravity 1.445
Molecular Formula C19H13NaO5S
Color Red
Synonym Phenolsulfonphthalein Sodium Salt
Quantity 5 g
Physical Form Solid
Molecular Weight (g/mol) 376.3583
Grade ACS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.