Learn More
Abcam Phenol red sodium salt, pH indicator, 25G
Catalog No. 502704706
Encompass_Preferred
MW 376.37 g/mol. Water soluble pH indicator used in the 6.8 (yellow) to 8.2 (red) range. Commonly employed as a cell tissue culture pH marker. Under normal conditions, phenol red added to the growth medium will have a pink-red color. Dying cells or overgrowth of contaminants will cause a change in pH resulting in a change in indicator color.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4706 Supplier Abcam Supplier No. AB14633725G
MW 376.37 g/mol. Water soluble pH indicator used in the 6.8 (yellow) to 8.2 (red) range. Commonly employed as a cell tissue culture pH marker. Under normal conditions, phenol red added to the growth medium will have a pink-red color. Dying cells or overgrowth of contaminants will cause a change in pH resulting in a change in indicator color.
The product is subject to the following: Abcam Restricted Use Statement