Learn More
Sigma Aldrich Fine Chemicals Biosciences Phenol Red sodium salt50G
Catalog No. 502897161
Encompass
Phenol Red sodium salt is widely used as a pH indicator. At a pH of 6.4 and below phenol red SOLUTIONon will have a yellow color. At a pH of 8.2 and above it will have a red color. It is commonly used in culture media to detect neutral to acidic pH changes.
Catalog No. 50-289-7161 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P475850G
Phenol Red sodium salt is widely used as a pH indicator. At a pH of 6.4 and below phenol red SOLUTIONon will have a yellow color. At a pH of 8.2 and above it will have a red color. It is commonly used in culture media to detect neutral to acidic pH changes.