Learn More
Hach Company Phenol Red Indicator Solution, Spec. Grade, 50 mL
Supplier:  Hach Company 2657512
Encompass
Phenol Red Indicator Solution. Standardized for colorimetric pH determination. Color range: pH 6.0-8.5. Bottle/50 mL spec. grade.
Catalog No. 50-199-6458