Learn More
Sigma Aldrich Fine Chemicals Biosciences Phenol solution, 108-95-2, MFCD00002143, 100mL
Catalog No. NC1641058
Encompass
Empirical Formula (Hill Notation) C6H6O, Molecular Weight 94.11, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology.
Catalog No. NC1641058 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P4557100ML
Empirical Formula (Hill Notation) C6H6O, Molecular Weight 94.11, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology.