Learn More
Sigma Aldrich Fine Chemicals Biosciences Phenol solution | 108-95-2 | MFCD00002143 | 400mL
Catalog No. 501884094
Encompass
Phenol solution | Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA | MW:94.11 | 108-95-2 | MFCD00002143 | 400mL
Catalog No. 50-188-4094 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P4557400ML
Phenol solution | Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA | MW:94.11 | 108-95-2 | MFCD00002143 | 400mL