Learn More
Thermo Scientific Chemicals Phenol/TRIS saturated sol., for molecular biology, stabilized, DNAse, RNAse and Protease free
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Phenol/Tris is a solvent system consisting of phenol and Tris(Hydroxymethyl)aminomethane
- Prepared as ready-to-use, no titration necessary
Applications
- Commonly used in DNA isolation. Tris(Hydroxymethyl)aminomethane acts as a proton acceptor to raise pH
Specifications
Specifications
| Chemical Name or Material | Phenol/TRIS saturated sol. |
| Color | Yellow to Orange |
| Name Note | Stabilized, DNase, RNase and Protease Free |
| CAS | 108-95-2, 77-86-1, 7732-18-5 |
| Composition | Phenol (108-95-2); Tris (hydroxymethyl) aminomethane (77-86-1) |
| Density | 1.07 |
| Molecular Formula | C6H6O |
| pH | 7.8 to 8.2 (25°C) |
| Quantity | 500 mL |
| MDL Number | MFCD01632719 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.