Learn More
Sigma Aldrich Fine Chemicals Biosciences PhenolChloroformIso-amyl
Catalog No. 502904921
Encompass
PhenolChloroformIso-amyl alcohol (125241) is used for the purification of total RNA from mixtures of crude cell extracts and can eliminate the need for a time consuming ultracentrifugation step.
Catalog No. 50-290-4921 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P1944400ML
PhenolChloroformIso-amyl alcohol (125241) is used for the purification of total RNA from mixtures of crude cell extracts and can eliminate the need for a time consuming ultracentrifugation step.