Learn More
Mark V Laboratory Inc Green Phenolic Mounting Powder, 5 LBS.
Catalog No. NC0925721
Encompass
This item has a minimum order quantity of 3 per supplier requirements Colored Phenolic Thermoset Powders are used in Mounting Press\"s for specimen mounting. Mounts are made at 150 degrees C at pressures between 3000-4200 psi. The different colors offer contrast to the parts being mounted when examined under a microscope.
Catalog No. NC0925721 Supplier Mark V Laboratory Inc Supplier No. MPG
This item has a minimum order quantity of 3 per supplier requirements Colored Phenolic Thermoset Powders are used in Mounting Press\"s for specimen mounting. Mounts are made at 150 degrees C at pressures between 3000-4200 psi. The different colors offer contrast to the parts being mounted when examined under a microscope.