Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Phenolic Screw Cap with Open Center

Catalog No. 111011883 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
240 mL (61 x 143 mm)
4 mL Vial, 13-425 Thread
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-101-1883 4 mL Vial, 13-425 Thread
11-101-1221 240 mL (61 x 143 mm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-1883 Supplier MilliporeSigma™ Supelco™ Supplier No. 27120U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenolic screw cap with open center

  • Available as hole caps for use with sealing septa
  • Phenolic hole caps can withstand a maximum temperature of 149°C.
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic
No. per Pack 100
Temperature Range (Metric) Temperature limit 149°C
Quantity 100 Pk.
For Use With (Equipment) 4 mL Vial, 13-425 Thread
Product Type Phenolic Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.