Learn More
Grainger Phenolic Tread Wheel 8 1400 lb.
Catalog No. 502766784
Encompass_Preferred
The Phenolic Tread Wheel is an 8 inch diameter wheel made of phenolic resin with a ribbed tread design. It has a 1/2 inch bore size and is rated for a maximum temperature of 250 degrees Fahrenheit. The wheel has a load capacity of 250 pounds per wheel and is designed for light duty applications like material handling carts, workbenches, and conveyors. The phenolic material provides good chemical resistance. Overall, this is a moderately rated phenolic wheel for light industrial uses requiring chemical resistance and heat tolerance up to 250°F.
Catalog No. 50-276-6784 Supplier Grainger Supplier No. 16V370
The Phenolic Tread Wheel is an 8 inch diameter wheel made of phenolic resin with a ribbed tread design. It has a 1/2 inch bore size and is rated for a maximum temperature of 250 degrees Fahrenheit. The wheel has a load capacity of 250 pounds per wheel and is designed for light duty applications like material handling carts, workbenches, and conveyors. The phenolic material provides good chemical resistance. Overall, this is a moderately rated phenolic wheel for light industrial uses requiring chemical resistance and heat tolerance up to 250°F.