Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spex CertiPrep™ Phenolics Mix
SDP

Catalog No. CLPSAIPK Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
4/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLPSAIPK 4/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLPSAIPK Supplier Spex CertiPrep™ Supplier No. CLPSAIPK
Add to Cart
Add to Cart

Analytical standard for municipal water, industrial water, and waste samples

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 604, 625, 8270 and CLP Series Methods

Specifications

Chemical Name or Material 2,4,6-Trichlorophenol
Packaging Bottle
CAS 88-06-2
Concentration 2,000 μg/mL
Color Clear
Melting Point Undetermined
Boiling Point 82°C (179.6°F)
Solution Type Standard Solution
Type Organic Pollutant Standard
Solvent or Matrix Methylene Chloride
Industry Type Environmental Testing
Quantity 4/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.