Learn More
Aqua Solutions Phenolphthalein 1% in 95% Alcohol (pH 8.0 Clear, pH 10.0 Pink) (1L)
SDP
Catalog No. NC9260104
Encompass_Preferred
Phenolphthalein 1% in 95% Alcohol (pH 8.0 Clear, pH 10.0 Pink) (1L)
Catalog No. NC9260104 Supplier Aqua Solutions Supplier No. P19971L
Phenolphthalein 1% in 95% Alcohol (pH 8.0 Clear, pH 10.0 Pink) (1L)