Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein, 1% (w/v) Indicator Solution in 70% Alcohol, Spectrum™ Chemical
SDP

Catalog No. 18612265 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-612-265 500 mL
18-612-266 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-612-265 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P153500MLPL
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Phenolphthalein
CAS 77-09-8, 64-17-5, 7732-18-5
Concentration 1%
Grade Chemical Solution
Packaging Poly Bottle
Color Colorless
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.