Learn More
Aqua Solutions Phenolphthalein 1% in Reagent Alcohol pH 8.0 Clear pH 10.0 Pink (500ML)
SDP
Catalog No. NC9815967
Encompass
Phenolphthalein 1% in Reagent Alcohol pH 8.0 Clear pH 10.0 Pink (500ML)
Catalog No. NC9815967 Supplier Aqua Solutions Supplier No. ACB037
Phenolphthalein 1% in Reagent Alcohol pH 8.0 Clear pH 10.0 Pink (500ML)