Learn More
Medchemexpress LLC HY-D0211 1g Medchemexpress, Phenolphthalein CAS:77-09-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD02111G
Encompass_Preferred
Medchemexpress, HY-D0211 1g Phenolphthalein CAS:77-09-8 Phenolphthalein is a widely applied but toxic indicator dye. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3158