Learn More
Aqua Solutions Phenolphthalein 0.5% w/v in Ethanol pH 8.0 Clear pH 10.0 Pink (ASTM 847) (100ML)
SDP
Catalog No. NC9009211
Encompass
Phenolphthalein 0.5% w/v in Ethanol pH 8.0 Clear pH 10.0 Pink (ASTM 847) (100ML)
Catalog No. NC9009211 Supplier Aqua Solutions Supplier No. TEN216
Phenolphthalein 0.5% w/v in Ethanol pH 8.0 Clear pH 10.0 Pink (ASTM 847) (100ML)