Learn More
Hach Company Phenolphthalein Indicator Solution, 5 g/L, 100 mL MDB (APHA)
Catalog No. 501994184 Shop All Hach Company Products
Encompass
pH indicator for use in titrations. Color change: colorless to pink at ~pH 8.3. 100 mL Marked Dropping Bottle.
Catalog No. 50-199-4184 Supplier Hach Company Supplier No. 16232
pH indicator for use in titrations. Color change: colorless to pink at ~pH 8.3. 100 mL Marked Dropping Bottle.