Learn More
Medchemexpress LLC Phenolphthalein | 77-09-8 | 99.0% | 25 G
SDP
Catalog No. 5000414371
Encompass_Preferred
Phenolphthalein is an acid-base indicator that remains colorless in an acidic environment and turns pink in an alkaline environment. It has been identified as genotoxic and potentially carcinogenic to mice. Additionally, phenolphthalein serves as a blood indicator.
  • Functions as an acid-base indicator.
  • Appears colorless in acidic conditions.
  • Turns pink in alkaline conditions.
  • Used as a blood indicator.
Catalog No. 50-004-14371 Supplier Medchemexpress LLC Supplier No. 25GHYD0211
Phenolphthalein is an acid-base indicator that remains colorless in an acidic environment and turns pink in an alkaline environment. It has been identified as genotoxic and potentially carcinogenic to mice. Additionally, phenolphthalein serves as a blood indicator.
  • Functions as an acid-base indicator.
  • Appears colorless in acidic conditions.
  • Turns pink in alkaline conditions.
  • Used as a blood indicator.