Learn More
Medchemexpress LLC Phenolphthalein | 77-09-8 | 99.0% | 50 G
SDP
Catalog No. 5000409808
Encompass_Preferred
Phenolphthalein is an acid-base indicator that is colorless in an acidic environment and pink in an alkaline environment. It is genotoxic and potentially carcinogenic to mice. Additionally, phenolphthalein is also a blood indicator. This product is for research use only.
  • Functions as an acid-base indicator, appearing colorless in acidic conditions and pink in alkaline conditions.
  • Noted as genotoxic and potentially carcinogenic to mice.
  • Also recognized as a blood indicator.
  • Data sheet, COA, SDS, and handling instructions are available for this product.
  • Soluble in DMSO at a concentration of ≥ 50 mg/mL (157.07 mM), with a note to use newly opened DMSO due to the impact of hygroscopic DMSO on solubility.
  • Recommended storage is at 4°C, protected from light. When in solvent, store at -80°C for 6 months or -20°C for 1 month, also protected from light.
Catalog No. 50-004-09808 Supplier Medchemexpress LLC Supplier No. HYD021150G
Phenolphthalein is an acid-base indicator that is colorless in an acidic environment and pink in an alkaline environment. It is genotoxic and potentially carcinogenic to mice. Additionally, phenolphthalein is also a blood indicator. This product is for research use only.
  • Functions as an acid-base indicator, appearing colorless in acidic conditions and pink in alkaline conditions.
  • Noted as genotoxic and potentially carcinogenic to mice.
  • Also recognized as a blood indicator.
  • Data sheet, COA, SDS, and handling instructions are available for this product.
  • Soluble in DMSO at a concentration of ≥ 50 mg/mL (157.07 mM), with a note to use newly opened DMSO due to the impact of hygroscopic DMSO on solubility.
  • Recommended storage is at 4°C, protected from light. When in solvent, store at -80°C for 6 months or -20°C for 1 month, also protected from light.