Learn More
Cayman Chemical PhenolphthaleIn DGlucuronI 5mg
SDP
Catalog No. 502990841
Encompass_Preferred
A colorimetric β-glucuronidase substrate; upon enzymatic cleavage by β-glucuronidase, phenolphthalein is released, which can be quantified by colorimetric detection at 552 nm as a measure of β-galactosidase activity
Catalog No. 50-299-0841 Supplier Cayman Chemical Supplier No. 304245 MG
A colorimetric β-glucuronidase substrate; upon enzymatic cleavage by β-glucuronidase, phenolphthalein is released, which can be quantified by colorimetric detection at 552 nm as a measure of β-galactosidase activity