Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein Diphosphate, Tetrasodium Salt Dihydrate 99%, Thermo Scientific™

Catalog No. AC227260050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC227260050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC227260050 Supplier Thermo Scientific Chemicals Supplier No. 227260050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68807-90-9
Molecular Formula C20H16Na4O10P2
Molecular Weight (g/mol) 570.245
InChI Key OXOCDDRINVOKHT-UHFFFAOYSA-N
PubChem CID 124203466
IUPAC Name [4-[3-oxo-1-(4-phosphonooxyphenyl)-2-benzofuran-1-yl]phenyl] dihydrogen phosphate;sodium
SMILES C1=CC=C2C(=C1)C(=O)OC2(C3=CC=C(C=C3)OP(=O)(O)O)C4=CC=C(C=C4)OP(=O)(O)O.[Na].[Na].[Na].[Na]

Specifications

Infrared Spectrum Authentic
Formula Weight 566.21
Percent Purity ≥98.5%
Color Orange/Pink
Quantity 5 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Phenolphthalein diphosphate, tetrasodium salt dihydrate, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.