Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenolphthalein glucuronide sodium | 6820-54-8 | 98.6% | 516.43 | 100 MG
SDP

Catalog No. 5000348525
Encompass_Preferred

Phenolphthalein glucuronide sodium is a biochemical assay reagent primarily utilized as a substrate for determining the activity of β-glucuronidase. This compound is offered with a high purity of 98.61% and appears as a solid with a color ranging from light yellow to brown. It is intended strictly for research purposes and is not for human use.

  • Acts as a substrate to quantify β-glucuronidase activity
  • High purity (98.6%)
  • Solid form, light yellow to brown in color
  • Molecular weight: 516.43
  • Store at -20°C under nitrogen, away from moisture; in solvent, store at -80°C for 6 months or -20°C for 1 month
  • Shipped at room temperature within the continental US; conditions may vary elsewhere
  • For research use only

Catalog No. 50-003-48525 Supplier Medchemexpress LLC Supplier No. HY137426A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenolphthalein glucuronide sodium is a biochemical assay reagent primarily utilized as a substrate for determining the activity of β-glucuronidase. This compound is offered with a high purity of 98.61% and appears as a solid with a color ranging from light yellow to brown. It is intended strictly for research purposes and is not for human use.

  • Acts as a substrate to quantify β-glucuronidase activity
  • High purity (98.6%)
  • Solid form, light yellow to brown in color
  • Molecular weight: 516.43
  • Store at -20°C under nitrogen, away from moisture; in solvent, store at -80°C for 6 months or -20°C for 1 month
  • Shipped at room temperature within the continental US; conditions may vary elsewhere
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.