Learn More
Medchemexpress LLC Phenolphthalein glucuronide sodium | 6820-54-8 | 98.6% | 516.43 | 50 MG
SDP
Catalog No. 5000349761
Encompass_Preferred
Phenolphthalein glucuronide sodium is a substrate used to determine β-glucuronidase activity. It is intended for research use only and is not sold to patients.
  • Substrate to determine β-glucuronidase activity
  • For research use only
Catalog No. 50-003-49761 Supplier Medchemexpress LLC Supplier No. HY137426A50MG
Phenolphthalein glucuronide sodium is a substrate used to determine β-glucuronidase activity. It is intended for research use only and is not sold to patients.
  • Substrate to determine β-glucuronidase activity
  • For research use only