Learn More
Medchemexpress LLC Phenolphthalein glucuronide sodium | 6820-54-8 | C26H21NaO10 | 10MG
SDP
Catalog No. 5000259028
Encompass_Preferred
Phenolphthalein glucuronide sodium is a chromogenic reagent used as a substrate for β-glucuronidase assays. Upon enzymatic hydrolysis it releases phenolphthalein, enabling sensitive colorimetric detection of β-glucuronidase activity in biochemical and enzymatic studies. Supplied for research use only.
Catalog No. 50-002-59028 Supplier Medchemexpress LLC Supplier No. HY137426A10MG
Phenolphthalein glucuronide sodium is a chromogenic reagent used as a substrate for β-glucuronidase assays. Upon enzymatic hydrolysis it releases phenolphthalein, enabling sensitive colorimetric detection of β-glucuronidase activity in biochemical and enzymatic studies. Supplied for research use only.