Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenolphthalein glucuronide sodium | 6820-54-8 | MFCD00051200 | >=90.0% | 516.43 g/mol | C26H21NaO10 | 5 MG
SDP

Catalog No. 5000222473
Encompass_Preferred

Phenolphthalein glucuronide sodium is the sodium salt of phenolphthalein glucuronide used as a chromogenic substrate to determine β-glucuronidase activity in biochemical assays. It is supplied as a solid and is provided with a product data sheet and handling instructions; storage recommendations are specified on the certificate of analysis.

  • Chromogenic substrate for β-glucuronidase assays.
  • Sodium salt supplied as a solid.
  • Molecular formula C26H21NaO10 and molecular weight 516.43 g/mol.
  • Available in multiple sizes from 5 mg to 500 mg.
  • Storage conditions specified on the certificate of analysis.
  • Data sheet and handling instructions available with the product.

Catalog No. 50-002-22473 Supplier Medchemexpress LLC Supplier No. HY137426A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenolphthalein glucuronide sodium is the sodium salt of phenolphthalein glucuronide used as a chromogenic substrate to determine β-glucuronidase activity in biochemical assays. It is supplied as a solid and is provided with a product data sheet and handling instructions; storage recommendations are specified on the certificate of analysis.

  • Chromogenic substrate for β-glucuronidase assays.
  • Sodium salt supplied as a solid.
  • Molecular formula C26H21NaO10 and molecular weight 516.43 g/mol.
  • Available in multiple sizes from 5 mg to 500 mg.
  • Storage conditions specified on the certificate of analysis.
  • Data sheet and handling instructions available with the product.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.