Learn More
Hach Company Phenolphthalein Indicator Solution, 100 mL
Supplier:  Hach Company 2330242
Encompass
pH indicator for use in titrations. Color change: colorless to pink at ~pH 8.3, 5 g/L in 50% isopropanol
Catalog No. 50-199-5821