Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein Indicator, 0.25% w/v in Methanol, Reagents

Catalog No. 01336185
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-185 500 mL
01-336-186 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-185 Supplier Reagents Holdings Llc Supplier No. C84003NE500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenolphthalein Indicator, 0.25% w/v in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Phenolphthalein Indicator, 0.25% w/v in Methanol
Boiling Point 65°C
CAS 67-56-1, 77-09-8
Packaging Poly Bottle
Specific Gravity 0.8
Molecular Formula C20H14O4/CH3OH
Color Colorless
Synonym 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
Quantity 500 mL
Physical Form Liquid
Percent Purity 0.25%
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.