Learn More
Phenolphthalein Indicator, 1% (w/v) in 70% (v/v) Alcohol, Neutralized, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Chemical Name or Material | Phenolphthalein Indicator |
| CAS | 64-17-5, 7732-18-5, 77-09-8, 67-56-1 |
| CAS Min % | 29.0, 65.0, 0.5, 2.0 |
| CAS Max % | 31.0, 69.0, 1.5, 4.0 |
| Concentration | 1% (w/v) |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | 70% (v/v) Alcohol |
| Color | Colorless to Pink |
| pH | 8 |
| Quantity | 120 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.