Learn More
Aqua Solutions Phenolphthalein 1% in 95% Alcohol (pH 8.0 Clear, pH 10.0 Pink) (100ML)
SDP
Catalog No. NC9681284
Encompass_Preferred
Phenolphthalein 1% in 95% Alcohol (pH 8.0 Clear, pH 10.0 Pink) (100ML)
Catalog No. NC9681284 Supplier Aqua Solutions Supplier No. P1997100ML
Phenolphthalein 1% in 95% Alcohol (pH 8.0 Clear, pH 10.0 Pink) (100ML)