Learn More
Description
- Changes from colorless to pink at ∼8.3 pH
- Compatible with commercially available titration kits
Specifications
Specifications
| pH Range | 0 to 14 |
| Volume (Metric) | 100 mL |
| For Use With (Application) | Calibration and Maintenance |
| For Use With (Equipment) | pH Meter and Electrode |
| Type | pH indicator |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.