Description
- Quantity
- 60 tests per kit
- Performance
- Simple, 4 step test
- Easy to use
- Immediate pink color on end of swab indicates a presumptive positive for blood
- Options and Accessories
- Phenolphthalein Presumptive Pocket Blood Test Kit (FS752)
- Phenolphthalein Pocket Refill (FS753)
Specifications
Specifications
| Product Type | Blood Test Kit |
| Packaging Type | Refill Pack |