Learn More
Hach Company Phenolphthalein Indicator Powder Pillows, pk/100
Supplier:  Hach Company 94299
Encompass
pH indicator for use in titrations. Color change: colorless to pink at ~pH 8.3. Pack of 100 powder pillows
Catalog No. 50-199-4473