Learn More
Hach Company Phenolphthalein Indicator Solution, 1 g/L, 15 mL
Supplier:  Hach Company 189736
Encompass
pH indicator for use in titrations. Color change: colorless to pink at ~pH 8.3
Catalog No. 50-199-4634