Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein Solution R 0.1% w/v in 80% v/v Alcohol, Reagents

Catalog No. 01334384
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-384 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-384 Supplier Reagents Holdings Llc Supplier No. CP108900500A
Add to Cart
Add to Cart

Phenolphthalein Solution R 0.1% w/v in 80% v/v Alcohol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Phenolphthalein Solution R 0.1% w/v in 80% v/v Alcohol
CAS 64-17-5, 7732-18-5, 77-09-8
Synonym 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.7893
Color Colorless
Quantity 500 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.