Learn More
Description
Phenolphthalein is an acid base indicator. It is a weak acid, which is colorless in acidic solution and turns puce pink in alkaline solution. Phenolphthalein is genotoxic. It might cause oxidative damage and binds to estrogen receptors.
Specifications
Specifications
| Chemical Name or Material | Phenolphthalein |
| CAS | 77-09-8 |
| Concentration | 0.5% |
| Color | Colorless |
| pH | 8 to 10 |
| Quantity | 100 mL |
| Molecular Weight (g/mol) | 318.32 g/mol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.